6-Bromo-5-fluoro-3-iodo-2-methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

473669-5G 473669-25G 473669-1G 473669-500MG
77629-686EA 1841.64 USD
77629-686 77635-918 77639-104 77652-882
6-Bromo-5-fluoro-3-iodo-2-methyl-2H-indazole ≥97%
6-Bromo-5-fluoro-3-iodo-2-methyl-2H-indazole

Precaución: For Research Use Only

Formula: C₈H₅BrFIN₂
Storage Temperature: Ambient
CAS Number: 3031789-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR