1-(2-Fluoro-3-(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

473622-5G 473622-1G 473622-500MG
77629-684EA 1484.23 USD
77629-684 77643-132 77651-096
1-(2-Fluoro-3-(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(2-Fluoro-3-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 222.179 g/mol
MW: C₁₀H₁₀F₄O
Storage Temperature: Ambient
CAS Number: 1504107-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR