Methyl 2-(2-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

473620-5G 473620-25G 473620-1G 473620-500MG
77629-682EA 1484.23 USD
77629-682 77635-916 77639-098 77651-094
Methyl 2-(2-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥97%
Methyl 2-(2-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 250.146 g/mol
MW: C₁₀H₆F₄O₃
Storage Temperature: Ambient
CAS Number: 3031804-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR