(2,6-Difluoro-4-iodophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

473601-5G 473601-1G 473601-25G 473601-500MG
77633-298EA 1441 USD
77633-298 77639-096 77659-590 77651-092
(2,6-Difluoro-4-iodophenyl)(phenyl)methanol ≥95%
(2,6-Difluoro-4-iodophenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₃H₉F₂IO
Storage Temperature: Ambient
CAS Number: 3037703-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR