1-(2,6-Difluoro-4-iodophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

473600-5G 473600-1G 473600-500MG 473600-25G
77629-680EA 1484.23 USD
77629-680 77639-094 77652-880 77656-012
1-(2,6-Difluoro-4-iodophenyl)propan-1-one ≥95%
1-(2,6-Difluoro-4-iodophenyl)propan-1-one

Precaución: For Research Use Only

Formula: 296.053 g/mol
MW: C₉H₇F₂IO
Storage Temperature: Ambient
CAS Number: 3037692-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR