N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide ≥95%

Supplier: AOBChem USA

Warning

Synonyms: N-(1-Hydroxy-2-methylpropan-2-yl)-2-methoxybenzamide

473397-5G 473397-25G 473397-1G 473397-500MG
77635-052EA 418 USD
77635-052 77656-010 77643-130 77647-108
N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide ≥95%
N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide

Precaución: For Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665208
CAS Number: 74201-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR