1-(Benzo[d]thiazol-2-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

473377-5G 473377-1G 473377-25G 473377-500MG
77633-296EA 1037 USD
77633-296 77639-090 77656-008 77651-090
1-(Benzo[d]thiazol-2-yl)-2,2,2-trifluoroethanone ≥97%
1-(Benzo[d]thiazol-2-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 231.1943 g/mol
MW: C₉H₄F₃NOS
Storage Temperature: Ambient
CAS Number: 929973-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR