5-((5-Bromopyridin-2-yl)oxy)-2,5-dimethylhexan-2-ol ≥95%

Supplier: AOBChem USA

Warning

473375-5G 473375-25G 473375-1G 473375-500MG
77629-676EA 1484.23 USD
77629-676 77635-912 77643-128 77651-088
5-((5-Bromopyridin-2-yl)oxy)-2,5-dimethylhexan-2-ol ≥95%
5-((5-Bromopyridin-2-yl)oxy)-2,5-dimethylhexan-2-ol

Precaución: For Research Use Only

Formula: 302.2074 g/mol
MW: C₁₃H₂₀BrNO₂
Storage Temperature: Ambient
CAS Number: 3037704-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR