3-(3-(Tert-butyl)phenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

473243-5G 473243-25G 473243-1G 473243-500MG
77629-674EA 1484.23 USD
77629-674 77656-006 77643-126 77647-106
3-(3-(Tert-butyl)phenyl)oxetan-3-ol ≥95%
3-(3-(Tert-butyl)phenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 206.281 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031812-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR