1-(3-(Tert-butyl)phenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

473242-1G 473242-5G 473242-500MG
77639-088EA 523 USD
77639-088 77659-588 77651-086
1-(3-(Tert-butyl)phenyl)-2-methoxyethanone ≥95%
1-(3-(Tert-butyl)phenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 206.281 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
CAS Number: 1256481-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR