1-(3-(Tert-butyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

473240-5G 473240-1G 473240-500MG
77659-586EA 1811 USD
77659-586 77643-124 77647-102
1-(3-(Tert-butyl)phenyl)cyclobutanol ≥95%
1-(3-(Tert-butyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₄H₂₀O
Storage Temperature: Ambient
CAS Number: 2092098-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR