1-(3-Ethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

473237-5G 473237-1G 473237-500MG
77656-002EA 1898 USD
77656-002 77643-120 77647-098
1-(3-Ethylphenyl)cyclobutanol ≥95%
1-(3-Ethylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
CAS Number: 1499855-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR