1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

473234-5G 473234-25G 473234-1G 473234-500MG
77629-670EA 1484.23 USD
77629-670 77659-582 77643-116 77647-096
1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)ethanol ≥95%
1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)ethanol

Precaución: For Research Use Only

Formula: 287.049 g/mol
MW: C₉H₇BrF₄O
Storage Temperature: Ambient
CAS Number: 3031788-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR