Tert-butyl 3-fluoro-5-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

473229-5G 473229-25G 473229-1G 473229-500MG
77633-292EA 668 USD
77633-292 77659-580 77643-114 77651-082
Tert-butyl 3-fluoro-5-(trifluoromethyl)benzoate ≥95%
Tert-butyl 3-fluoro-5-(trifluoromethyl)benzoate

Precaución: For Research Use Only

Formula: C₁₂H₁₂F₄O₂
MW: 264.216 g/mol
Storage Temperature: Ambient
CAS Number: 3031809-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR