(3-Bromo-2-fluoro-6-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

472872-5G 472872-25G 472872-1G 472872-500MG
77633-288EA 788 USD
77633-288 77659-576 77643-110 77647-094
(3-Bromo-2-fluoro-6-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%
(3-Bromo-2-fluoro-6-(trifluoromethoxy)phenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 305.087 g/mol
MW: C₈H₅BrF₄OS
Storage Temperature: Ambient
CAS Number: 2168222-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR