Cyclopropyl(2,5-dimethoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

472853-500MG 472853-5G 472853-1G
77652-878EA 400 USD
77652-878 77655-998 77643-108
Cyclopropyl(2,5-dimethoxyphenyl)methanone ≥95%
Cyclopropyl(2,5-dimethoxyphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₄O₃
Storage Temperature: Ambient
CAS Number: 120350-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR