2-(3-Fluoro-4-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

472844-5G 472844-1G 472844-500MG
77629-668EA 1484.23 USD
77629-668 77643-106 77647-090
2-(3-Fluoro-4-methylphenyl)propan-2-ol ≥95%
2-(3-Fluoro-4-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 168.208 g/mol
MW: C₁₀H₁₃FO
Storage Temperature: Ambient
CAS Number: 1379326-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR