2-Chloro-2-fluoro-1-(3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

472732-5G 472732-25G 472732-1G 472732-500MG
77629-666EA 1484.23 USD
77629-666 77659-574 77643-104 77651-078
2-Chloro-2-fluoro-1-(3-methylphenyl)ethanone ≥95%
2-Chloro-2-fluoro-1-(3-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 186.61 g/mol
MW: C₉H₈ClFO
Storage Temperature: Ambient
CAS Number: 2768979-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR