6-Hydroxy-4-methylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

47231-5G 47231-25G 47231-1G 47231-500MG
77633-284EA 1250 USD
77633-284 77655-996 77643-102 77647-088
6-Hydroxy-4-methylnicotinic acid ≥97%
6-Hydroxy-4-methylnicotinic acid

Precaución: For Research Use Only

Formula: C₇H₇NO₃
MW: 153.135 g/mol
Storage Temperature: Ambient
CAS Number: 66909-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR