2,4-Difluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47222-5G 47222-500MG 47222-25G 47222-1G
77635-050EA 578 USD
77635-050 77652-876 77655-994 77643-100
2,4-Difluoro-5-methylbenzaldehyde ≥97%
2,4-Difluoro-5-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD12196967
CAS Number: 315204-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR