2-Chloro-1-(3-methoxy-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

471585-5G 471585-1G 471585-500MG 471585-25G
77633-282EA 1171 USD
77633-282 77639-080 77652-874 77659-568
2-Chloro-1-(3-methoxy-4-methylphenyl)ethanone ≥95%
2-Chloro-1-(3-methoxy-4-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
CAS Number: 1378815-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR