2-Chloro-1-[2-fluoro-3-(trifluoromethyl)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

471532-5G 471532-1G 471532-500MG
77629-664EA 1484.23 USD
77629-664 77639-078 77651-074
2-Chloro-1-[2-fluoro-3-(trifluoromethyl)phenyl]ethanone ≥95%
2-Chloro-1-[2-fluoro-3-(trifluoromethyl)phenyl]ethanone

Precaución: For Research Use Only

Formula: 240.58 g/mol
MW: C₉H₅ClF₄O
Storage Temperature: Ambient
CAS Number: 881401-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR