2-Chloro-1-(5-methyl-2-pyridinyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

471528-1G 471528-500MG
77635-048EA 1473 USD
77635-048 77647-086
2-Chloro-1-(5-methyl-2-pyridinyl)ethanone ≥95%
2-Chloro-1-(5-methyl-2-pyridinyl)ethanone

Precaución: For Research Use Only

Formula: 169.61 g/mol
MW: C₈H₈ClNO
Storage Temperature: Ambient
CAS Number: 1379245-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR