Ethanone, 2-chloro-1-(2-fluoro-5-methylphenyl)- ≥95%

Supplier: AOBChem USA

Warning

471507-5G 471507-1G 471507-25G 471507-500MG
77633-280EA 1320 USD
77633-280 77639-076 77659-566 77651-072
Ethanone, 2-chloro-1-(2-fluoro-5-methylphenyl)- ≥95%
Ethanone, 2-chloro-1-(2-fluoro-5-methylphenyl)-

Precaución: For Research Use Only

Formula: 186.61 g/mol
MW: C₉H₈ClFO
Storage Temperature: Ambient
CAS Number: 66122-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR