2-Chloro-1-(2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

471384-5G 471384-500MG 471384-25G 471384-1G
77633-276EA 750 USD
77633-276 77652-872 77655-990 77643-096
2-Chloro-1-(2-fluorophenyl)ethanone ≥95%
2-Chloro-1-(2-fluorophenyl)ethanone

Precaución: For Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD13194396
CAS Number: 53688-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR