α-3-Cyclohexen-1-yl-3-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

470569-5G 470569-25G 470569-1G 470569-500MG
77629-662EA 1484.23 USD
77629-662 77635-910 77639-072 77647-082
α-3-Cyclohexen-1-yl-3-methoxybenzenemethanol ≥95%
α-3-Cyclohexen-1-yl-3-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: 218.29 g/mol
MW: C₁₄H₁₈O₂
Storage Temperature: Ambient
CAS Number: 1342070-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR