4-Fluoro-2-iodobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

46988-5G 46988-1G 46988-25G 46988-500MG
77635-046EA 429 USD
77635-046 77639-070 77659-560 77647-080
4-Fluoro-2-iodobenzaldehyde ≥95%
4-Fluoro-2-iodobenzaldehyde

Precaución: For Research Use Only

Formula: 250.009 g/mol
MW: C₇H₄FIO
Storage Temperature: Ambient
MDL Number: MFCD12025051
CAS Number: 909545-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR