2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

469302-5G 469302-1G 469302-25G 469302-500MG
77629-660EA 1484.23 USD
77629-660 77639-066 77659-554 77651-064
2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propan-2-ol ≥95%
2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 230.2774 g/mol
MW: C₁₅H₁₅FO
Storage Temperature: Ambient
CAS Number: 55258-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR