(5-Chloro-4-methylpyridin-3-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

469075-5G 469075-1G 469075-25G 469075-500MG
77629-658EA 1484.23 USD
77629-658 77639-064 77655-988 77651-062
(5-Chloro-4-methylpyridin-3-yl)(phenyl)methanol ≥95%
(5-Chloro-4-methylpyridin-3-yl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 233.693 g/mol
MW: C₁₃H₁₂ClNO
Storage Temperature: Ambient
CAS Number: 3031813-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR