1-(3-Ethoxy-4-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

469044-5G 469044-25G 469044-1G 469044-500MG
77629-654EA 1484.23 USD
77629-654 77655-986 77643-092 77651-060
1-(3-Ethoxy-4-methylphenyl)cyclopentanol ≥95%
1-(3-Ethoxy-4-methylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 220.307 g/mol
MW: C₁₄H₂₀O₂
Storage Temperature: Ambient
CAS Number: 3031798-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR