1-(3-Ethoxy-4-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

469023-5G 469023-1G 469023-25G 469023-500MG
77629-648EA 1553.24 USD
77629-648 77639-058 77655-978 77647-064
1-(3-Ethoxy-4-methylphenyl)propan-1-one ≥95%
1-(3-Ethoxy-4-methylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 192.254 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
CAS Number: 1806614-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR