2-Chloro-1-(3-ethoxy-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

469022-5G 469022-1G 469022-25G 469022-500MG
77629-646EA 1484.23 USD
77629-646 77639-056 77655-976 77647-062
2-Chloro-1-(3-ethoxy-4-methylphenyl)ethanone ≥95%
2-Chloro-1-(3-ethoxy-4-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 212.673 g/mol
MW: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
CAS Number: 3037704-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR