Ethyl 2-(6-(cyclohexyloxy)pyridin-3-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

468940-5G 468940-25G 468940-1G 468940-500MG
77629-644EA 1676.84 USD
77629-644 77635-908 77643-088 77651-056
Ethyl 2-(6-(cyclohexyloxy)pyridin-3-yl)-2-oxoacetate ≥95%
Ethyl 2-(6-(cyclohexyloxy)pyridin-3-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₅H₁₉NO₄
Storage Temperature: Ambient
CAS Number: 3037682-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR