1-(6-(Cyclohexyloxy)pyridin-3-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

468938-5G 468938-500MG 468938-25G 468938-1G
77629-642EA 1851.94 USD
77629-642 77652-870 77659-548 77643-086
1-(6-(Cyclohexyloxy)pyridin-3-yl)-2,2,2-trifluoroethanone ≥97%
1-(6-(Cyclohexyloxy)pyridin-3-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 273.251 g/mol
MW: C₁₃H₁₄F₃NO₂
Storage Temperature: Ambient
CAS Number: 3037682-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR