2-(Cyclohexyloxy)-5-(trimethylsilyl)pyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
468935-5G
468935-25G
468935-1G
468935-500MG
77629-638EA
1484.23
USD
77629-638
77655-972
77647-056
77647-058
2-(Cyclohexyloxy)-5-(trimethylsilyl)pyridine ≥95%
2-(Cyclohexyloxy)-5-(trimethylsilyl)pyridine
Precaución: For Research Use Only
Formula:
249.424 g/mol MW: C₁₄H₂₃NOSi Storage Temperature: Ambient |
CAS Number:
3037676-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...