1-(5-Cyclohexyl-2-methoxyphenyl)-2-fluoroethanone ≥97%

Supplier: AOBChem USA

Warning

468917-5G 468917-25G 468917-1G 468917-500MG
77629-636EA 1484.23 USD
77629-636 77655-970 77643-084 77647-054
1-(5-Cyclohexyl-2-methoxyphenyl)-2-fluoroethanone ≥97%
1-(5-Cyclohexyl-2-methoxyphenyl)-2-fluoroethanone

Precaución: For Research Use Only

Formula: C₁₅H₁₉FO₂
Storage Temperature: Ambient
CAS Number: 3037703-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR