Ethyl 2-(5-cyclohexyl-2-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

468912-5G 468912-25G 468912-1G 468912-500MG
77633-266EA 1288 USD
77633-266 77655-968 77643-082 77651-054
Ethyl 2-(5-cyclohexyl-2-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(5-cyclohexyl-2-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₇H₂₂O₄
Storage Temperature: Ambient
CAS Number: 1892247-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR