(5-Cyclohexyl-2-methoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

468906-5G 468906-1G 468906-25G 468906-500MG
77629-632EA 1583.11 USD
77629-632 77639-052 77659-546 77651-050
(5-Cyclohexyl-2-methoxyphenyl)trimethylsilane ≥95%
(5-Cyclohexyl-2-methoxyphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 262.463 g/mol
MW: C₁₆H₂₆OSi
Storage Temperature: Ambient
CAS Number: 3037704-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR