Ethyl 2-(3-cyclopentyl-4-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

468119-5G 468119-25G 468119-1G 468119-500MG
77629-628EA 1484.23 USD
77629-628 77635-906 77639-044 77651-042
Ethyl 2-(3-cyclopentyl-4-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-cyclopentyl-4-fluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₅H₁₇FO₃
Storage Temperature: Ambient
CAS Number: 3037692-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR