2-(3-Benzyl-5-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

468113-1G 468113-5G 468113-500MG
77639-040EA 696 USD
77639-040 77659-540 77651-038
2-(3-Benzyl-5-methylphenyl)propan-2-ol ≥95%
2-(3-Benzyl-5-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 240.34 g/mol
MW: C₁₇H₂₀O
Storage Temperature: Ambient
CAS Number: 3037696-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR