1-(3-Phenoxyphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

468106-5G 468106-25G 468106-1G 468106-500MG
77629-616EA 1484.23 USD
77629-616 77655-954 77643-070 77647-040
1-(3-Phenoxyphenyl)propan-1-ol ≥95%
1-(3-Phenoxyphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 228.286 g/mol
MW: C₁₅H₁₆O₂
Storage Temperature: Ambient
CAS Number: 63403-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR