2-(3-Phenoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

468105-5G 468105-1G 468105-500MG
77629-614EA 1484.23 USD
77629-614 77639-038 77651-036
2-(3-Phenoxyphenyl)propan-2-ol ≥95%
2-(3-Phenoxyphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 228.286 g/mol
MW: C₁₅H₁₆O₂
Storage Temperature: Ambient
CAS Number: 2153578-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR