2-Chloro-1-(4-ethoxy-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

468098-5G 468098-500MG 468098-25G 468098-1G
77629-610EA 1484.23 USD
77629-610 77652-866 77659-536 77643-066
2-Chloro-1-(4-ethoxy-2-fluorophenyl)ethanone ≥95%
2-Chloro-1-(4-ethoxy-2-fluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 216.637 g/mol
MW: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
CAS Number: 3037703-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR