1-(4-Ethoxy-2-fluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

468097-5G 468097-1G 468097-25G 468097-500MG
77633-260EA 1206 USD
77633-260 77639-036 77655-952 77647-038
1-(4-Ethoxy-2-fluorophenyl)propan-1-one ≥95%
1-(4-Ethoxy-2-fluorophenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
CAS Number: 1805894-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR