2-(4-Ethoxy-2-fluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

468096-1G 468096-5G 468096-500MG
77639-034EA 666 USD
77639-034 77655-950 77647-036
2-(4-Ethoxy-2-fluorophenyl)propan-2-ol ≥95%
2-(4-Ethoxy-2-fluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₅FO₂
Storage Temperature: Ambient
CAS Number: 1780492-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR