Tert-butyl 4-ethoxy-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

468095-5G 468095-25G 468095-1G 468095-500MG
77629-608EA 1484.23 USD
77629-608 77635-900 77643-064 77647-034
Tert-butyl 4-ethoxy-2-fluorobenzoate ≥97%
Tert-butyl 4-ethoxy-2-fluorobenzoate

Precaución: For Research Use Only

Formula: C₁₃H₁₇FO₃
Storage Temperature: Ambient
CAS Number: 3037693-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR