1-Ethoxy-4-iodo-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

468079-5G 468079-1G 468079-25G 468079-500MG
77629-600EA 1484.23 USD
77629-600 77639-028 77659-526 77651-030
1-Ethoxy-4-iodo-2-(trifluoromethyl)benzene ≥97%
1-Ethoxy-4-iodo-2-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 316.059 g/mol
MW: C₉H₈F₃IO
Storage Temperature: Ambient
CAS Number: 2384552-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR