2-(3-Ethoxy-4-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

468054-5G 468054-25G 468054-1G 468054-500MG
77629-598EA 1484.23 USD
77629-598 77659-524 77643-056 77651-028
2-(3-Ethoxy-4-methylphenyl)propan-2-ol ≥95%
2-(3-Ethoxy-4-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031799-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR