Tert-butyl 3-ethoxy-4-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

468053-5G 468053-25G 468053-1G 468053-500MG
77629-596EA 1612.98 USD
77629-596 77635-896 77639-026 77647-026
Tert-butyl 3-ethoxy-4-methylbenzoate ≥95%
Tert-butyl 3-ethoxy-4-methylbenzoate

Precaución: For Research Use Only

Formula: 236.307 g/mol
MW: C₁₄H₂₀O₃
Storage Temperature: Ambient
CAS Number: 3031789-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR