2-Ethoxy-4-iodo-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

468050-1G 468050-5G 468050-500MG
77639-022EA 531 USD
77639-022 77659-522 77651-024
2-Ethoxy-4-iodo-1-methylbenzene ≥95%
2-Ethoxy-4-iodo-1-methylbenzene

Precaución: For Research Use Only

Formula: 262.088 g/mol
MW: C₉H₁₁IO
Storage Temperature: Ambient
CAS Number: 1855795-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR